accessibility menu, dialog, popup

UserName

CAS RN 6005-99-8

IUPAC Name: 1-phenyltridecan-1-one
Synonyms: 1-phenyltridecan-1-one
Molecular Weight (g/mol): 274.45
Molecular Formula: C19H30O
InChi Key: ZZNDQCACFUJAKJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results