missing translation for 'onlineSavingsMsg'
Learn More

Tridecanenitrile 96.0+%, TCI America™

Catalog No. T081925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T081925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T081925G Supplier TCI America Supplier No. T081925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-60-7
Molecular Formula C13H25N
Molecular Weight (g/mol) 195.35
MDL Number MFCD00039492
InChI Key WKJHMKQSIBMURP-UHFFFAOYSA-N
Synonym dodecyl cyanide, 1-cyanododecane, n-dodecyl cyanide, n-tridecanonitrile, tridecane nitrile, nitriles, c10-16, c10-c16 alkylnitrile, tridecannitril, tridecanonitrile, lauryl cyanide
PubChem CID 12390
IUPAC Name tridecanenitrile
SMILES CCCCCCCCCCCCC#N

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 195.35
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecanenitrile
EINECSNumber (2)-1507
TSCA Yes