CAS RN 629-60-7
IUPAC Name:
tridecanenitrile
Synonyms:
tridecanenitrile
Molecular Weight (g/mol):
195.35
Molecular Formula:
C13H25N
InChi Key:
WKJHMKQSIBMURP-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCC#N
1
–
1
of
1
results
1
Tridecanenitrile 96.0+%, TCI America™
CAS: 629-60-7 Molecular Formula: C13H25N Molecular Weight (g/mol): 195.35 MDL Number: MFCD00039492 InChI Key: WKJHMKQSIBMURP-UHFFFAOYSA-N Synonym: dodecyl cyanide,1-cyanododecane,n-dodecyl cyanide,n-tridecanonitrile,tridecane nitrile,nitriles, c10-16,c10-c16 alkylnitrile,tridecannitril,tridecanonitrile,lauryl cyanide PubChem CID: 12390 IUPAC Name: tridecanenitrile SMILES: CCCCCCCCCCCCC#N