missing translation for 'onlineSavingsMsg'
Learn More

Tridecanedioic Acid 96.0+%, TCI America™

Supplier:  TCI America T002125G

 View more versions of this product

Catalog No. T002125G


Only null left

Description

Description

Specifications

Specifications

Tridecanedioic Acid
113°C
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
C(CCCCCC(=O)O)CCCCCC(=O)O
244.331
CHEBI:73718
≥96.0% (GC,T)
505-52-2
C13H24O4
25 g
DXNCZXXFRKPEPY-UHFFFAOYSA-N
tridecanedioic acid
10458
244.33
Crystalline Powder

Chemical Identifiers

505-52-2
244.331
DXNCZXXFRKPEPY-UHFFFAOYSA-N
10458
tridecanedioic acid
C13H24O4
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
CHEBI:73718
C(CCCCCC(=O)O)CCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0878

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions