missing translation for 'onlineSavingsMsg'
Learn More

Tridecanedioic Acid 96.0+%, TCI America™

$271.51 - $271.51

Chemical Identifiers

CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.331
MDL Number MFCD00002740
InChI Key DXNCZXXFRKPEPY-UHFFFAOYSA-N
Synonym brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
PubChem CID 10458
ChEBI CHEBI:73718
IUPAC Name tridecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T002125G
View Documents
TCI America
T002125G
25 g
Each for $271.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

505-52-2
244.331
DXNCZXXFRKPEPY-UHFFFAOYSA-N
10458
tridecanedioic acid
C13H24O4
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
CHEBI:73718
C(CCCCCC(=O)O)CCCCCC(=O)O

Specifications

505-52-2
C13H24O4
25 g
DXNCZXXFRKPEPY-UHFFFAOYSA-N
tridecanedioic acid
10458
244.33
Crystalline Powder
113°C
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
C(CCCCCC(=O)O)CCCCCC(=O)O
244.331
CHEBI:73718
≥96.0% (GC,T)
Tridecanedioic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0878

TSCA : Yes

SDS
Documents

Documents