missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tridecane | |
| -5°C | |
| 235°C | |
| MFCD00008979 | |
| n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
| CCCCCCCCCCCCC | |
| 184.37 | |
| CHEBI:35998 | |
| ≥99.0% (GC) |
| 629-50-5 | |
| Colorless | |
| C13H28 | |
| 500 mL | |
| IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
| tridecane | |
| 12388 | |
| 184.37 | |
| Liquid |
Chemical Identifiers
| 629-50-5 | |
| 184.37 | |
| IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
| 12388 | |
| tridecane |
| C13H28 | |
| MFCD00008979 | |
| n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
| CHEBI:35998 | |
| CCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0010
RTECSNumber : YD3025000
TSCA : Yes