missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tridecane 99.0+%, TCI America™
$57.79 - $160.30
Chemical Identifiers
| CAS | 629-50-5 |
|---|---|
| Molecular Formula | C13H28 |
| Molecular Weight (g/mol) | 184.37 |
| MDL Number | MFCD00008979 |
| InChI Key | IIYFAKIEWZDVMP-UHFFFAOYSA-N |
| Synonym | n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group |
| PubChem CID | 12388 |
| ChEBI | CHEBI:35998 |
| IUPAC Name | tridecane |
| SMILES | CCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T041125ML
|
TCI America
T041125ML |
25 mL |
Each for $57.79
|
|
|||||
|
T0411100ML
|
TCI America
T0411100ML |
100 mL |
Each for $115.57
|
|
|||||
|
T0411500ML
|
TCI America
T0411500ML |
500 mL |
Each for $160.30
|
|
|||||
Chemical Identifiers
| 629-50-5 | |
| 184.37 | |
| IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
| 12388 | |
| tridecane |
| C13H28 | |
| MFCD00008979 | |
| n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
| CHEBI:35998 | |
| CCCCCCCCCCCCC |
Specifications
| 629-50-5 | |
| Colorless | |
| C13H28 | |
| 25 mL | |
| IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
| tridecane | |
| 12388 | |
| 184.37 | |
| Liquid |
| -5°C | |
| 235°C | |
| MFCD00008979 | |
| n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
| CCCCCCCCCCCCC | |
| 184.37 | |
| CHEBI:35998 | |
| ≥99.0% (GC) | |
| Tridecane |
Safety and Handling
EINECSNumber : (2)-0010
RTECSNumber : YD3025000
TSCA : Yes