missing translation for 'onlineSavingsMsg'
Learn More

Tridecan-7-amine 97.0+%, TCI America™

Catalog No. p-7137806
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35911ML 1 mL
T35915ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35911ML Supplier TCI America Supplier No. T35911ML

Chemical Identifiers

CAS 22513-16-2
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
MDL Number MFCD20690568
InChI Key FXNJNGMYIZRQRG-UHFFFAOYSA-N
Synonym 1-Hexylheptylamine, 7-Aminotridecane
PubChem CID 574991
IUPAC Name tridecan-7-amine
SMILES CCCCCCC(CCCCCC)N

Specifications

Color Colorless
Boiling Point 102°C
Quantity 1 mL
UN Number 2735
Formula Weight 199.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecan-7-amine
EINECSNumber (2)-0133&(2)-0176
TSCA No