accessibility menu, dialog, popup

UserName

H3CCH3NH2

CAS RN 22513-16-2

IUPAC Name: tridecan-7-amine
Synonyms: tridecan-7-amine
Molecular Weight (g/mol): 199.38
Molecular Formula: C13H29N
InChi Key: FXNJNGMYIZRQRG-UHFFFAOYSA-N
SMILES: CCCCCCC(N)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results