missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7137297
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25851G 1 g
T25855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25851G Supplier TCI America Supplier No. T25851G
Only null left

Chemical Identifiers

CAS 58656-04-5
Molecular Formula C18H34BF4P
Molecular Weight (g/mol) 368.25
MDL Number MFCD03840580
InChI Key MYSMMEUXKHJYKH-UHFFFAOYSA-O
Synonym tricyclohexylphosphonium tetrafluoroborate, tricyclohexylphosphine tetrafluoroborate, tricyclohexylphosphine tetrafluroborate, tricyclohexylphosphoniumtetrafluoroborate, pcy3.hbf4, ksc269e4n, tricyclohexylphosphine tetrofluroborate, tricyclohexyl phosphonium tetrafluoroborate, tetrafluoroboranuide tricyclohexylphosphanium, tricyclohexyl-phosphonium tetrafluoro borate
PubChem CID 11710396
IUPAC Name tetrafluoroboranuide; tricyclohexylphosphanium
SMILES F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1

Specifications

Melting Point 170°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 368.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tricyclohexylphosphonium Tetrafluoroborate