accessibility menu, dialog, popup

UserName

BFFFFPH

CAS RN 58656-04-5

IUPAC Name: tetrafluoroboranuide; tricyclohexylphosphanium
Synonyms: tricyclohexylphosphanium tetrafluoroborate
Molecular Weight (g/mol): 368.25
Molecular Formula: C18H34BF4P
InChi Key: MYSMMEUXKHJYKH-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)