missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphonium tetrafluoroborate, 99%

Catalog No. p-3523611
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC386830010 1 g Glass bottle
AC386830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC386830010 Supplier Thermo Scientific Chemicals Supplier No. 386830010

CAS: 58656-04-5 | C18H34BF4P | 368.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58656-04-5
Molecular Formula C18H34BF4P
Molecular Weight (g/mol) 368.25
MDL Number MFCD03840580
InChI Key MYSMMEUXKHJYKH-UHFFFAOYSA-O
Synonym tricyclohexylphosphonium tetrafluoroborate, tricyclohexylphosphine tetrafluoroborate, tricyclohexylphosphine tetrafluroborate, tricyclohexylphosphoniumtetrafluoroborate, pcy3.hbf4, ksc269e4n, tricyclohexylphosphine tetrofluroborate, tricyclohexyl phosphonium tetrafluoroborate, tetrafluoroboranuide tricyclohexylphosphanium, tricyclohexyl-phosphonium tetrafluoro borate
PubChem CID 11710396
SMILES F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1

Specifications

Melting Point 165.0°C to 169.0°C
Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
IUPAC Name tricyclohexylphosphanium;tetrafluoroborate
Formula Weight 368.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only