missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™, Thermo Scientific™

Catalog No. AC428421000
Change view
Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428421000 100mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428421000 Supplier Thermo Scientific Chemicals Supplier No. 428421000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2622-14-2 , 108-88-3
Molecular Formula C18H33P
Molecular Weight (g/mol) 280.43
MDL Number MFCD00003853
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
PubChem CID 75806
IUPAC Name tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

Specifications

Density 0.9010g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Quantity 100mL
Specific Gravity 0.901
Solubility Information Solubility in water: insoluble
Formula Weight 280.43
Chemical Name or Material Tricyclohexylphosphine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May cause respiratory irritation. Suspected of damaging fertility or the unborn child. Harmful if swallowed. Causes serious eye irritation. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
EINECSNumber 220-069-2

RUO – Research Use Only