missing translation for 'onlineSavingsMsg'
Learn More

(Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate, 99%

Catalog No. AC363581000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363581000 100 mg Glass bottle
AC363585000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC363581000 Supplier Thermo Scientific Chemicals Supplier No. 363581000

CAS: 64536-78-3 | C31H50F6IrNP2 | 804.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64536-78-3
Molecular Formula C31H50F6IrNP2
Molecular Weight (g/mol) 804.90
MDL Number MFCD00075097
InChI Key UJXHUUQZACSUOG-KJWGIZLLSA-N
Synonym crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate
PubChem CID 5702647
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;pyridine;tricyclohexylphosphane;hexafluorophosphate
SMILES [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 175.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 mg
Fieser 10,116; 12,151; 13,88
Merck Index 15, 2554
Solubility Information Solubility in water: insoluble
Formula Weight 804.89
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (Tricyclohexylphosphine)(1, 5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only