accessibility menu, dialog, popup

UserName

CAS RN 64536-78-3

IUPAC Name: iridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide pyridine tricyclohexylphosphane
Synonyms: crabtree's catalyst
Molecular Weight (g/mol): 804.9
Molecular Formula: C31H50F6IrNP2
InChi Key: UJXHUUQZACSUOG-KJWGIZLLSA-N
SMILES: [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)