missing translation for 'onlineSavingsMsg'
Learn More

Tricyclo[6.2.1.0(2,7)]undeca-4-ene 96.0+%, TCI America™

Catalog No. T1384500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1384500ML 500 mL
T138425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1384500ML Supplier TCI America Supplier No. T1384500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91465-71-3
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00153968
InChI Key YPOHZFOZYNRWKX-UHFFFAOYSA-N
PubChem CID 548751
SMILES C1CC2CC1C3C2CC=CC3

Specifications

Color Yellow
Boiling Point 209°C
Quantity 500 mL
Formula Weight 148.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tricyclo[6.2.1.0(2,7)]undeca-4-ene