accessibility menu, dialog, popup

UserName

CAS RN 91465-71-3

IUPAC Name: tricyclo[6.2.1.0²,⁷]undec-4-ene
Synonyms: tricyclo[6.2.1.0²,⁷]undec-4-ene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: YPOHZFOZYNRWKX-UHFFFAOYNA-N
SMILES: C1CC2CC1C1CC=CCC21
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results