missing translation for 'onlineSavingsMsg'
Learn More

Tricosanoic Acid 95.0+%, TCI America™

Supplier:  TCI America T040210G

 View more versions of this product

Catalog No. T040210G


Only null left

Description

Description

Specifications

Specifications

Tricosanoic Acid
80°C
C23H46O2
10 g
XEZVDURJDFGERA-UHFFFAOYSA-N
tricosanoic acid
17085
354.62
Crystalline Powder
2433-96-7
White-Yellow
MFCD00002808
n-tricosanoic acid, unii-muc9a0ms6v, 1-tricosanoic acid, n-tricosanoic, muc9a0ms6v, tricosanoicacid, tricosylic acid, n-tricosanoate, acmc-1cr37, 94087-10-2 aluminum salt
CCCCCCCCCCCCCCCCCCCCCCC(=O)O
354.619
CHEBI:42394
≥95.0% (GC,T)

Chemical Identifiers

2433-96-7
354.619
XEZVDURJDFGERA-UHFFFAOYSA-N
17085
tricosanoic acid
C23H46O2
MFCD00002808
n-tricosanoic acid, unii-muc9a0ms6v, 1-tricosanoic acid, n-tricosanoic, muc9a0ms6v, tricosanoicacid, tricosylic acid, n-tricosanoate, acmc-1cr37, 94087-10-2 aluminum salt
CHEBI:42394
CCCCCCCCCCCCCCCCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : No

SDS
Documents

Documents

Promotions

Promotions