accessibility menu, dialog, popup

UserName

CAS RN 2433-96-7

H3COHO

CAS RN 2433-96-7

IUPAC Name: tricosanoic acid
Synonyms: tricosanoic acid
Molecular Weight (g/mol): 354.62
Molecular Formula: C23H46O2
InChi Key: XEZVDURJDFGERA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results