missing translation for 'onlineSavingsMsg'
Learn More

Tricosanoic Acid 95.0+%, TCI America™

Catalog No. T04021G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04021G 1 g
T040210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04021G Supplier TCI America Supplier No. T04021G
Only null left

Chemical Identifiers

CAS 2433-96-7
Molecular Formula C23H46O2
Molecular Weight (g/mol) 354.619
MDL Number MFCD00002808
InChI Key XEZVDURJDFGERA-UHFFFAOYSA-N
Synonym n-tricosanoic acid, unii-muc9a0ms6v, 1-tricosanoic acid, n-tricosanoic, muc9a0ms6v, tricosanoicacid, tricosylic acid, n-tricosanoate, acmc-1cr37, 94087-10-2 aluminum salt
PubChem CID 17085
ChEBI CHEBI:42394
IUPAC Name tricosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
Formula Weight 354.62
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tricosanoic Acid
EINECSNumber (2)-0608
TSCA No