missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triclabendazole 98.0+%, TCI America™
Supplier: TCI America T28261G
Specifications
| Triclabendazole | |
| 176°C | |
| MFCD00864519 | |
| triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole | |
| CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl | |
| 359.649 | |
| 359.65 | |
| Crystalline Powder |
| 68786-66-3 | |
| C14H9Cl3N2OS | |
| 1 g | |
| NQPDXQQQCQDHHW-UHFFFAOYSA-N | |
| 6-chloro-5-(2,3-dichlorophenoxy)-2-methylsulfanyl-1H-benzimidazole | |
| 50248 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 68786-66-3 | |
| 359.649 | |
| NQPDXQQQCQDHHW-UHFFFAOYSA-N | |
| 50248 | |
| CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl |
| C14H9Cl3N2OS | |
| MFCD00864519 | |
| triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole | |
| 6-chloro-5-(2,3-dichlorophenoxy)-2-methylsulfanyl-1H-benzimidazole |
Safety and Handling
RTECSNumber : DD6747000
TSCA : No