missing translation for 'onlineSavingsMsg'
Learn More

Trichostatin A 98.0+%, TCI America™

Catalog No. p-7137234
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T247710MG 10 mg
T247750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T247710MG Supplier TCI America Supplier No. T247710MG

Chemical Identifiers

CAS 58880-19-6
Molecular Formula C17H22N2O3
Molecular Weight (g/mol) 302.37
MDL Number MFCD03848392
InChI Key RTKIYFITIVXBLE-WKWSCTOINA-N
Synonym trichostatin a, trichostatin a tsa, trichostatin, tsa, unii-3x2s926l3z, 2e,4e,6r-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, gnf-pf-1011, 2,4-heptadienamide, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide, r-e,e-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
PubChem CID 444732
ChEBI CHEBI:46024
IUPAC Name (2E,4E)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
SMILES CC(\C=C(/C)\C=C\C(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 10 mg
Formula Weight 302.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trichostatin A
RTECSNumber MI5215000
TSCA No
Recommended Storage Freezer