missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricholine Citrate (ca. 65% in Water), TCI America™
Supplier: TCI America C033025G
Specifications
| Tricholine Citrate (ca. 65% in Water) | |
| C21H47N3O10 | |
| 25 g | |
| RPERJPYDELTDMR-UHFFFAOYSA-K | |
| 2-hydroxyethyl(trimethyl)azanium;2-hydroxypropane-1,2,3-tricarboxylate | |
| 11024 |
| 546-63-4 | |
| MFCD00059985 | |
| Choline Citrate Solution | |
| C[N+](C)(C)CCO.C[N+](C)(C)CCO.C[N+](C)(C)CCO.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O | |
| 501.618 | |
| Liquid |
Chemical Identifiers
| 546-63-4 | |
| 501.618 | |
| RPERJPYDELTDMR-UHFFFAOYSA-K | |
| 11024 | |
| C[N+](C)(C)CCO.C[N+](C)(C)CCO.C[N+](C)(C)CCO.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O |
| C21H47N3O10 | |
| MFCD00059985 | |
| Choline Citrate Solution | |
| 2-hydroxyethyl(trimethyl)azanium;2-hydroxypropane-1,2,3-tricarboxylate |
Safety and Handling
TSCA : Yes