missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichlorovinylsilane 98.0+%, TCI America™
$44.73 - $268.43
Chemical Identifiers
| CAS | 75-94-5 |
|---|---|
| Molecular Formula | C2H3Cl3Si |
| Molecular Weight (g/mol) | 161.481 |
| MDL Number | MFCD00000482 |
| InChI Key | GQIUQDDJKHLHTB-UHFFFAOYSA-N |
| Synonym | trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride |
| PubChem CID | 6413 |
| IUPAC Name | trichloro(ethenyl)silane |
| SMILES | C=C[Si](Cl)(Cl)Cl |
Chemical Identifiers
| 75-94-5 | |
| 161.481 | |
| GQIUQDDJKHLHTB-UHFFFAOYSA-N | |
| 6413 | |
| C=C[Si](Cl)(Cl)Cl |
| C2H3Cl3Si | |
| MFCD00000482 | |
| trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride | |
| trichloro(ethenyl)silane |
Specifications
| 75-94-5 | |
| 92°C | |
| MFCD00000482 | |
| 1305 | |
| GQIUQDDJKHLHTB-UHFFFAOYSA-N | |
| trichloro(ethenyl)silane | |
| 6413 | |
| ≥98.0% (GC) | |
| Trichlorovinylsilane |
| Yellow | |
| C2H3Cl3Si | |
| 25 g | |
| trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride | |
| C=C[Si](Cl)(Cl)Cl | |
| 161.481 | |
| 161.48 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2037
RTECSNumber : VV6125000
TSCA : Yes