CAS RN 75-94-5
CAS RN 75-94-5
IUPAC Name:
trichloro(ethenyl)silane
Synonyms:
vinyltrichlorosilane
Molecular Weight (g/mol):
161.48
Molecular Formula:
C2H3Cl3Si
InChi Key:
GQIUQDDJKHLHTB-UHFFFAOYSA-N
SMILES:
Cl[Si](Cl)(Cl)C=C
1
–
1
of
1
results
1
Trichlorovinylsilane 98.0+%, TCI America™
CAS: 75-94-5 Molecular Formula: C2H3Cl3Si Molecular Weight (g/mol): 161.481 MDL Number: MFCD00000482 InChI Key: GQIUQDDJKHLHTB-UHFFFAOYSA-N Synonym: trichlorovinylsilane,vinyltrichlorosilane,trichloro vinyl silane,vtcs,silane, trichloroethenyl,silane, trichlorovinyl,trichlorovinylsilicon,vinyl trichlorosilane,trichlorovinyl silicane,vinylsilicon trichloride PubChem CID: 6413 IUPAC Name: trichloro(ethenyl)silane SMILES: C=C[Si](Cl)(Cl)Cl