missing translation for 'onlineSavingsMsg'
Learn More

Trichlorocyclopentylsilane 95.0+%, TCI America™

Catalog No. T25765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25765G 5 g
T257625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25765G Supplier TCI America Supplier No. T25765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14579-03-4
Molecular Formula C5H9Cl3Si
Molecular Weight (g/mol) 203.562
MDL Number MFCD00236508
InChI Key FCMZRNUHEXJWGB-UHFFFAOYSA-N
Synonym cyclopentyltrichlorosilane, trichloro cyclopentyl silane, silane, trichlorocyclopentyl, cyclopentane, trichlorosilyl, trichlorosilylcyclopentane, cyclopentyl trichlorosilane, acmc-1c6jw, trichlorocyclopentylsilane
PubChem CID 84533
IUPAC Name trichloro(cyclopentyl)silane
SMILES C1CCC(C1)[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 178°C
Quantity 5 g
UN Number 2987
Formula Weight 203.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichlorocyclopentylsilane