accessibility menu, dialog, popup

UserName

CAS RN 14579-03-4

IUPAC Name: trichloro(cyclopentyl)silane
Synonyms: trichloro(cyclopentyl)silane
Molecular Weight (g/mol): 203.56
Molecular Formula: C5H9Cl3Si
InChi Key: FCMZRNUHEXJWGB-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)C1CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results