missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, sodium salt, 97%

Catalog No. AC184151000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184151000 100 g Plastic bottle
AC184155000 500 g Plastic bottle
AC184150025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC184151000 Supplier Thermo Scientific Chemicals Supplier No. 184151000

CAS: 650-51-1 | C2Cl3NaO2 | 185.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 650-51-1
Molecular Formula C2Cl3NaO2
Molecular Weight (g/mol) 185.36
MDL Number MFCD00064198
InChI Key SAQSTQBVENFSKT-UHFFFAOYSA-M
Synonym sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca
PubChem CID 23681045
ChEBI CHEBI:81976
IUPAC Name sodium;2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)[O-].[Na+]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Plastic bottle
Linear Formula CCl3CO2Na
Quantity 100 g
Fieser 01,1107; 02,388; 06,553; 07,341
Merck Index 15, 9792
Solubility Information Solubility in water: soluble (153 g/100mL at 20°C). Other solubilities: soluble in ethanol, acetone, diethyl ether
Formula Weight 185.37
Percent Purity 0.97
Physical Form Powder or Granular Powder
Chemical Name or Material Trichloroacetic acid, sodium salt
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-479-2
RTECSNumber AJ9100000
TSCA TSCA

RUO – Research Use Only