missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Fisher Chemical

Catalog No. SA433500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA433500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA433500 Supplier Thermo Fisher Scientific Supplier No. SA433 500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-03-9 , 7732-18-5
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Melting Point 54°C
Boiling Point 196°C
Color Colorless
Density 1.62g/cm³
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5 to 101.0 %
UN Number 2564
Formula Weight 163.39
Percent Purity 99.5 to 101.0%
Concentration or Composition (by Analyte or Components) 6.12M (100% w/v)
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive