Learn More
Applied Biosystems™ Trichloroacetic Acid Dichloromethane, TCA DCM Solution
Catalog No. p-7083033
Encompass
Change view
Click to view available options
Quantity:
2 L
4 x 2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4347377 4 x 2 L
401272 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4347377 Supplier Applied Biosystems™ Supplier No. 4347377
Only null left
This Trichloroacetic acid (TCA)/Dichloromethane (DCM) solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.
Specifications
Quantity 4 x 2 L
For Research Use Only. Not for use in diagnostic procedures.