missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, Certified, 80.0% (w/v) ±2.0%, LabChem™
Specifications
Specifications
| Color | Colorless to Tan |
| CAS Max % | 80, 20 |
| Quantity | 500 mL |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Traceability to NIST | Traceable to NIST |
| Linear Formula | C2HCl3O2 |
| UN Number | UN2564 |
| Solubility Information | Soluble in water |
| Formula Weight | 163.39 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Suspected of causing cancer (oral). |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |