missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. p-5388964
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC262301 500 mL
LC262302 1 L
LC262304 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC262301 Supplier LabChem Supplier No. LC262301
Only null left

Chemical Identifiers

CAS 76-03-9 , 7732-18-5
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color Colorless
CAS Max % 90, 10
Linear Formula C2HCl3O2
UN Number UN2564
Solubility Information Soluble in water
Formula Weight 163.39
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Quantity 500 mL
Packaging Amber Glass
Physical Form Liquid
Traceability to NIST Traceable to NIST
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer (oral).
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature