Learn More
Trichloroacetic Acid, Certified, 10.0% (w/v) ±0.5%, LabChem™
Supplier: LabChem LC262302
Specifications
| Trichloroacetic Acid | |
| 90,10 | |
| C2HCl3O2 | |
| UN2564 | |
| Soluble in water | |
| C(=O)(C(Cl)(Cl)Cl)O | |
| 163.378 | |
| CHEBI:30956 | |
| 10.0% ±0.5% (w/v) | |
| 1 L | |
| Liquid |
| 76-03-9,7732-18-5 | |
| Colorless | |
| C2HCl3O2 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 2,2,2-trichloroacetic acid | |
| 6421 | |
| 163.39 | |
| Certified | |
| Amber Glass | |
| Traceable to NIST |
Chemical Identifiers
| 76-03-9 | |
| 163.378 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| CHEBI:30956 | |
| C(=O)(C(Cl)(Cl)Cl)O |
| C2HCl3O2 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 6421 | |
| 2,2,2-trichloroacetic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer (oral).
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Danger
Recommended Storage : Room Temperature