missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 50% (w/v) Aqueous Solution, Spectrum™ Chemical

Catalog No. 18614516
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614516 4 L Amber Glass Bottle
18614514 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614516 Supplier Spectrum Chemical Mfg Cor Supplier No. T1824LTGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-03-9 , 7732-18-5

Specifications

Quantity 4 L
Packaging Amber Glass Bottle
Percent Purity 50% ±1.5%
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.