missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 5% (w/v), Ricca Chemical

Catalog No. p-2917144
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
86804 120 mL Amber Glass Bottle
868016 500 mL Amber Glass Bottle
868032 1 L Amber Glass Bottle
3 options

Catalog No. 86804

Supplier: Ricca Chemical Company RI86804

Only null left

Chemical Identifiers

CAS 7732-18-5 , 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) Mixture
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

Percent Purity 5% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Quantity 120 mL
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic Acid