missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, 20% (w/v), Ricca Chemical
$175.75 - $175.75
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C2HCl3O2 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
| Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
| PubChem CID | 6421 |
| ChEBI | CHEBI:30956 |
| IUPAC Name | 2,2,2-trichloroacetic acid |
| SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| CHEBI:30956 | |
| C(=O)(C(Cl)(Cl)Cl)O |
| C2HCl3O2 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 6421 | |
| 2,2,2-trichloroacetic acid |
Specifications
| 7732-18-5,76-03-9 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| C(=O)(C(Cl)(Cl)Cl)O | |
| Mixture | |
| CHEBI:30956 | |
| Laboratory | |
| Amber Glass Bottle |
| C2HCl3O2 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 2,2,2-trichloroacetic acid | |
| 6421 | |
| 20% w/v | |
| 500 mL | |
| Trichloroacetic acid |