missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 15% (w/v), Ricca Chemical

Catalog No. 13622625
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13622625 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13622625 Supplier Ricca Chemical Company Supplier No. 8686.51

Chemical Identifiers

CAS 7732-18-5 , 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) Mixture
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Percent Purity 15% w/v
Grade Laboratory
Quantity 4 L
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic acid