missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 10% (w/v), Ricca Chemical

Catalog No. 86854
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
86854 120 mL Amber Glass Bottle
868516 500 mL Amber Glass Bottle
868532 1 L Amber Glass Bottle
86851 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 86854 Supplier Ricca Chemical Company Supplier No. RI86854

Chemical Identifiers

CAS 7732-18-5 , 76-03-9 , 7440-44-0
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) Mixture
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

CAS Min % 8.9, 89.0, 0.18
CAS Max % 9.26, 93.0, 0.19
Concentration 10% (w/v)
Percent Purity 10% w/v
Grade Laboratory
Quantity 120 mL
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic Acid