missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 868516
Specifications
| Trichloroacetic Acid | |
| 8.9,89.0,0.18 | |
| C2HCl3O2 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 10% (w/v) | |
| Mixture | |
| CHEBI:30956 | |
| Laboratory | |
| Amber Glass Bottle |
| 7732-18-5,76-03-9,7440-44-0 | |
| 9.26,93.0,0.19 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| OC(=O)C(Cl)(Cl)Cl | |
| trichloroacetic acid | |
| 6421 | |
| 10% w/v | |
| 500 mL |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| CHEBI:30956 | |
| OC(=O)C(Cl)(Cl)Cl |
| C2HCl3O2 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 6421 | |
| trichloroacetic acid |