missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonyldichlororuthenium(II) dimer

Catalog No. AA1050303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1050303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1050303 Supplier Thermo Scientific Chemicals Supplier No. 01050303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22594-69-0 | C6Cl4O6Ru2 | 512.00 g/mol

Tricarbonyldichlororuthenium(II) dimer is used to enhance the coagulation and attenuation of vulnerability to fibrinolysis, modify thrombus growth or disintegration, enhances fibrinogen as a substrate for thrombin, and regulation of ion transport by gasotransmitters. Also used for various pharmacological studies. It is used as a CO donor for reactive oxygen species mediated bacterial killing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22594-69-0
Molecular Formula C6Cl4O6Ru2
Molecular Weight (g/mol) 512.00
MDL Number MFCD00011528
InChI Key JYHHJVKGDCZCCL-UHFFFAOYSA-J
Synonym corm-2, tricarbonyldichlororuthenium ii dimer, rucl2 co 3 2, dichlorotricarbonylruthenium ii dimer, carbon monooxide-dichlororuthenium 3/1
PubChem CID 10951331
IUPAC Name carbon monoxide;dichlororuthenium
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Quantity 1 g
UN Number UN3466
Solubility Information Insoluble in water
Formula Weight 512.02
Physical Form Crystalline
Chemical Name or Material Tricarbonyldichlororuthenium(II) dimer
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only