missing translation for 'onlineSavingsMsg'
Learn More

Tricaprylin 93.0+%, TCI America™

Catalog No. T036525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T036525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T036525ML Supplier TCI America Supplier No. T036525ML
Only null left

Chemical Identifiers

CAS 538-23-8
Molecular Formula C27H50O6
Molecular Weight (g/mol) 470.691
MDL Number MFCD00036236
InChI Key VLPFTAMPNXLGLX-UHFFFAOYSA-N
Synonym tricaprylin, trioctanoin, rato, glycerol tricaprylate, glycerol trioctanoate, tricaprilin, caprylin, trioctanoylglycerol, maceight, glyceryl trioctanoate
PubChem CID 10850
ChEBI CHEBI:76978
IUPAC Name 2,3-di(octanoyloxy)propyl octanoate
SMILES CCCCCCCC(=O)OCC(COC(=O)CCCCCCC)OC(=O)CCCCCCC

Specifications

Melting Point 10°C
Boiling Point 234°C
Quantity 25 mL
Formula Weight 470.69
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tricaprylin
EINECSNumber (2)-0666
RTECSNumber YJ7700000
TSCA Yes