accessibility menu, dialog, popup

UserName

CAS RN 538-23-8

H3COOOOCH3OOCH3

CAS RN 538-23-8

IUPAC Name: 1,3-bis(octanoyloxy)propan-2-yl octanoate
Synonyms: Rato maceight trioctonoin trioctanoylglycerol octanoin, tri- (8CI) glyceryl tricaprylate glyceryl trioctanoate sefsol 800 tricaprylin tricaprylic glyceride
Molecular Weight (g/mol): 470.69
Molecular Formula: C27H50O6
InChi Key: VLPFTAMPNXLGLX-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCC(COC(=O)CCCCCCC)OC(=O)CCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results