missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tributyrin | |
| -75°C | |
| 190°C | |
| MFCD00009392 | |
| tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
| 302.367 | |
| CHEBI:35020 | |
| ≥98.0% (GC) |
| 60-01-5 | |
| Yellow | |
| C15H26O6 | |
| 25 mL | |
| UYXTWWCETRIEDR-UHFFFAOYSA-N | |
| 2,3-di(butanoyloxy)propyl butanoate | |
| 6050 | |
| 302.37 | |
| Liquid |
Chemical Identifiers
| 60-01-5 | |
| 302.367 | |
| UYXTWWCETRIEDR-UHFFFAOYSA-N | |
| 6050 | |
| 2,3-di(butanoyloxy)propyl butanoate |
| C15H26O6 | |
| MFCD00009392 | |
| tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
| CHEBI:35020 | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Safety and Handling
EINECSNumber : (2)-0654&(2)-0666
RTECSNumber : ET7350000
TSCA : Yes