missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyrin 98.0+%, TCI America™
$49.36 - $349.34
Chemical Identifiers
| CAS | 60-01-5 |
|---|---|
| Molecular Formula | C15H26O6 |
| Molecular Weight (g/mol) | 302.367 |
| MDL Number | MFCD00009392 |
| InChI Key | UYXTWWCETRIEDR-UHFFFAOYSA-N |
| Synonym | tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin |
| PubChem CID | 6050 |
| ChEBI | CHEBI:35020 |
| IUPAC Name | 2,3-di(butanoyloxy)propyl butanoate |
| SMILES | CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
T036425ML
|
N/A | 25 mL |
Each for $49.36
|
|
|||||
|
T0364500ML
|
N/A | 500 mL |
Each for $349.34
|
|
|||||
Chemical Identifiers
| C15H26O6 | |
| MFCD00009392 | |
| tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
| CHEBI:35020 | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Specifications
| 60-01-5 | |
| Yellow | |
| C15H26O6 | |
| tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
| 302.367 | |
| CHEBI:35020 | |
| ≥98.0% (GC) | |
| Tributyrin |
| -75°C | |
| 190°C | |
| MFCD00009392 | |
| UYXTWWCETRIEDR-UHFFFAOYSA-N | |
| 2,3-di(butanoyloxy)propyl butanoate | |
| 6050 | |
| 302.37 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0654&(2)-0666
RTECSNumber : ET7350000
TSCA : Yes