missing translation for 'onlineSavingsMsg'
Learn More

Tributylphenyltin

Catalog No. p-2692376
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC370230010 1 g Glass bottle
AC370230100 10 g Glass bottle
AC370230500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC370230010 Supplier Thermo Scientific Chemicals Supplier No. 370230010
Only null left

CAS: 960-16-7 | C18H32Sn | 367.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 960-16-7
Molecular Formula C18H32Sn
Molecular Weight (g/mol) 367.16
MDL Number MFCD00134394
InChI Key SYUVAXDZVWPKSI-UHFFFAOYSA-N
Synonym tributyl phenyl stannane, tributylphenyltin, phenyltributylstannane, stannane, tributylphenyl, phenyltributyltin, phenyltri-n-butyltin, phenyltributlytin, tributylphenyl tin, phenyl tnbutyl tin, phenyl tributyl tin
PubChem CID 607632
IUPAC Name tributyl(phenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1

Specifications

Color Colorless
Density 1.1250g/mL
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5150 to 1.5170
Linear Formula C6H5Sn[(CH2)3CH3]3
Quantity 1 g
Specific Gravity 1.125
Formula Weight 367.16
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Tributylphenyltin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for se

RUO – Research Use Only