accessibility menu, dialog, popup

UserName

CAS RN 960-16-7

IUPAC Name: (2,3,4-tributylphenyl)stannanylidyne
Synonyms: (2,3,4-tributylphenyl)stannanylidyne
Molecular Weight (g/mol): 364.14
Molecular Formula: C18H29Sn
InChi Key: ZMUNITMSSGNSOT-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C([Sn])C(CCCC)=C1CCCC
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)