missing translation for 'onlineSavingsMsg'
Learn More

Tributylmethylphosphonium Iodide 98.0+%, TCI America™

Catalog No. p-7132745
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14555G 5 g
M145525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14555G Supplier TCI America Supplier No. M14555G

Chemical Identifiers

CAS 1702-42-7
Molecular Formula C13H30IP
Molecular Weight (g/mol) 344.261
MDL Number MFCD00031626
InChI Key RLZMYANQLOCZOB-UHFFFAOYSA-M
Synonym Methyltributylphosphonium Iodide
PubChem CID 2760114
IUPAC Name tributyl(methyl)phosphanium;iodide
SMILES CCCC[P+](C)(CCCC)CCCC.[I-]

Specifications

Quantity 5 g
Formula Weight 344.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tributylmethylphosphonium Iodide
EINECSNumber (2)-4016
TSCA No