accessibility menu, dialog, popup

UserName

IH3CPH3CH3CH3C

CAS RN 1702-42-7

IUPAC Name: tributyl(methyl)phosphanium iodide
Synonyms: tributyl(methyl)phosphanium iodide
Molecular Weight (g/mol): 344.26
Molecular Formula: C13H30IP
InChi Key: RLZMYANQLOCZOB-UHFFFAOYSA-M
SMILES: [I-].CCCC[P+](C)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results