missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylmethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
Supplier: TCI America T267925G
Specifications
| Tributylmethylammonium Bis(trifluoromethanesulfonyl)imide | |
| Yellow | |
| MFCD12761447 | |
| XALVHDZWUBSWES-UHFFFAOYSA-N | |
| tributyl(methyl)azanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane | |
| 44630019 | |
| ≥98.0% (T) |
| 405514-94-5 | |
| C15H30F6N2O4S2 | |
| 25 g | |
| CCCC[N+](C)(CCCC)CCCC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 480.53 | |
| 480.53 | |
| Liquid |
Chemical Identifiers
| 405514-94-5 | |
| 480.53 | |
| XALVHDZWUBSWES-UHFFFAOYSA-N | |
| tributyl(methyl)azanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane |
| C15H30F6N2O4S2 | |
| MFCD12761447 | |
| 44630019 | |
| CCCC[N+](C)(CCCC)CCCC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Safety and Handling
TSCA : No