accessibility menu, dialog, popup

UserName

CAS RN 405514-94-5

H3CNH3CH3CH3CFFFSOONSOOFFF

CAS RN 405514-94-5

IUPAC Name: tributyl(methyl)azanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; tributyl(methyl)azanium
Molecular Weight (g/mol): 480.53
Molecular Formula: C15H30F6N2O4S2
InChi Key: XALVHDZWUBSWES-UHFFFAOYSA-N
SMILES: CCCC[N+](C)(CCCC)CCCC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results